Assay Details
Target Gene Details
Entrez Gene ID: | 72058 |
Gene Name: | immunoglobulin superfamily, member 5 |
Gene Aliases: |
2010003D20Rik, JCAM, Jam4 |
Location: |
Chr.16:96360467-96465572 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf5 | NM_001177886.1 | 3 | 316 | NP_001171357.1 |
| NM_001177887.1 | 3 | 320 | NP_001171358.1 | |
| NM_028078.3 | 3 | 313 | NP_082354.1 | |
| AF537215.1 | 3 | 254 | AAP49218.1 | |
| AK008060.1 | 3 | 284 | BAB25436.1 | |
| AK078934.1 | 3 | 284 | BAC37469.1 | |
| AK086973.1 | 3 | 317 | ||
| BC004806.1 | 3 | 266 | AAH04806.1 | |
| BX511978.1 | 3 | 344 | ||
| BY033616.1 | 3 | 314 |
More Information
Set Membership: |
|

Genomic Map