Assay Details
Target Gene Details
Entrez Gene ID: | 12647 |
Gene Name: | choline acetyltransferase |
Gene Aliases: |
B230380D24Rik |
Location: |
Chr.14:32408203-32465909 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chat | NM_009891.2 | NP_034021.1 | ||
| AK046406.1 | BAC32710.1 | |||
| D12490.1 | BAA02057.1 | |||
| D12491.1 | BAA20978.1 | |||
| D12492.1 | BAA20979.1 | |||
| D12493.1 | BAA20980.1 |
Target Gene Details
Entrez Gene ID: | 20508 |
Gene Name: | solute carrier family 18 (vesicular monoamine), member 3 |
Gene Aliases: |
VAChT, VAT |
Location: |
Chr.14:32462437-32464850 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc18a3 | NM_021712.2 | 1 | 1610 | NP_068358.2 |
| AK158592.1 | 1 | 1611 | BAE34572.1 | |
| BC120498.1 | 1 | 1227 | ||
| BC137705.1 | 1 | 1226 |
More Information
Set Membership: |
|

Genomic Map