Assay Details
Target Gene Details
Entrez Gene ID: | 75288 |
Gene Name: | solute carrier family 35, member F4 |
Gene Aliases: |
4930550L21Rik |
Location: |
Chr.14:49298519-49525837 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35f4 | NM_029238.2 | 3 | 564 | NP_083514.1 |
| AB041538.1 | 3 | 592 | BAA95023.1 | |
| AK034308.1 | 3 | 566 | BAC28670.1 | |
| AK043617.1 | 3 | 582 | ||
| AK046093.1 | 3 | 774 | BAC32602.1 | |
| AK047949.1 | 3 | 561 | BAC33198.1 | |
| BC138981.1 | 3 | 463 | ||
| BC145720.1 | 3 | 463 |

Genomic Map