Assay Details
Target Gene Details
Entrez Gene ID: | 671535 |
Gene Name: | poly (ADP-ribose) polymerase family, member 10 |
Gene Aliases: |
- |
Location: |
Chr.15:76232995-76243440 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp10 | NM_001163575.1 | 5 | 1320 | NP_001157047.1 |
| NM_001163576.1 | 4 | 1424 | NP_001157048.1 | |
| AK028373.1 | 5 | 1268 | BAC25913.1 | |
| AK035309.1 | 4 | 1394 | ||
| AK046755.1 | 4 | 1405 | BAC32856.1 | |
| AK146336.1 | 5 | 1321 | ||
| AK153205.1 | 4 | 1161 | BAE31805.1 | |
| AK166291.1 | 5 | 1271 | BAE38685.1 | |
| BC023056.1 | 1 | 157 | AAH23056.1 | |
| BC024074.1 | 5 | 1277 | AAH24074.1 | |
| BC025608.1 | 1 | 302 | AAH25608.1 |

Genomic Map