Assay Details
Target Gene Details
Entrez Gene ID: | 13840 |
Gene Name: | Eph receptor A6 |
Gene Aliases: |
Ehk2, Hek12, m-ehk2 |
Location: |
Chr.16:59653061-60606484 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha6 | NM_007938.2 | 1 | 117 | NP_031964.2 |
| AK090366.1 | 1 | 118 | ||
| AK135075.1 | 1 | 101 |
More Information
Set Membership: |
|

Genomic Map