Assay Details
Target Gene Details
Entrez Gene ID: | 68646 |
Gene Name: | NAD kinase 2, mitochondrial |
Gene Aliases: |
MNADK, Nadkd1 |
Location: |
Chr.15:9071260-9110496 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nadk2 | NM_001040395.4 | 1 | 168 | NP_001035485.2 |
NM_001085410.2 | 1 | 168 | NP_001078879.1 | |
NM_001286253.1 | 1 | 168 | NP_001273182.1 | |
NM_001286255.1 | 1 | 168 | NP_001273184.1 | |
AK046666.1 | 1 | 169 |
