Assay Details
Target Gene Details
Entrez Gene ID: | 223776 |
Gene Name: | selenoprotein O |
Gene Aliases: |
1300018J18Rik |
Location: |
Chr.15:89089107-89100340 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selo | NM_027905.2 | 5 | 1127 | NP_082181.2 |
| AK005048.1 | 5 | 1127 | BAB23774.1 | |
| AK054432.1 | 5 | 1089 | ||
| AK088096.1 | 5 | 1094 | ||
| AK161059.1 | 5 | 1078 | ||
| BC048849.1 | 2 | 150 | AAH48849.1 | |
| BC117817.1 | 6 | 642 | ||
| BC141293.1 | 5 | 1196 |

Genomic Map