Assay Details
Target Gene Details
Entrez Gene ID: | 66656 |
Gene Name: | eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein) |
Gene Aliases: |
EF-1-delta |
Location: |
Chr.15:75894796-75909556 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef1d | NM_001285429.1 | 8 | 1307 | NP_001272358.1 |
| NM_001285430.1 | 8 | 1307 | NP_001272359.1 | |
| NM_001285431.1 | 7 | 1235 | NP_001272360.1 | |
| NM_001285432.1 | 7 | 1238 | NP_001272361.1 | |
| NM_001285433.1 | 7 | 1235 | NP_001272362.1 | |
| NM_001285434.1 | 8 | 1292 | NP_001272363.1 | |
| NM_023240.3 | 8 | 1310 | NP_075729.2 | |
| NM_029663.2 | 10 | 2455 | NP_083939.1 | |
| AK017616.1 | 7 | 1190 | BAB30841.1 | |
| AK017796.1 | 8 | 1268 |
More Information
Set Membership: |
|

Genomic Map