Assay Details
Target Gene Details
Entrez Gene ID: | 20512 |
Gene Name: | solute carrier family 1 (glial high affinity glutamate transporter), member 3 |
Gene Aliases: |
AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1 |
Location: |
Chr.15:8634124-8710807 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc1a3 | NM_148938.3 | 6 | 1189 | NP_683740.1 |
| AF330257.1 | 5 | 689 | AAK01708.1 | |
| AK031831.1 | 6 | 852 | ||
| AK040798.1 | 6 | 849 | BAC30705.1 | |
| AK045949.1 | 6 | 868 | BAC32544.1 | |
| AK049090.1 | 6 | 849 | BAC33538.1 | |
| AK080879.1 | 6 | 873 | ||
| AK082736.1 | 6 | 849 | BAC38592.1 | |
| AK084968.1 | 6 | 833 | BAC39324.1 | |
| AK158777.1 | 5 | 723 | BAE34658.1 | |
| BC058711.1 | 6 | 1112 | AAH58711.1 | |
| BC066154.1 | 6 | 792 | AAH66154.1 | |
| L19565.1 | 3 | 172 | AAK13573.1 |
More Information
Set Membership: |
|

Genomic Map