Assay Details
Target Gene Details
Entrez Gene ID: | 212514 |
Gene Name: | spindle and centriole associated protein 1 |
Gene Aliases: |
Ccdc52, D16Ertd480e |
Location: |
Chr.16:44347395-44388492 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spice1 | NM_144550.4 | 14 | 1938 | NP_653133.3 |
AK048789.1 | 14 | 1882 | BAC33458.1 | |
AY099108.1 | 14 | 2218 | AAM34496.1 | |
BC052381.1 | 14 | 1892 | AAH52381.1 | |
BC056960.1 | 14 | 1892 | AAH56960.1 |
