Assay Details
Target Gene Details
Entrez Gene ID: | 238988 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 2 |
Gene Aliases: |
6430531D06, CAST, CAST1/ERC2, D14Ertd171e, ELKS2alpha |
Location: |
Chr.14:27622410-28478537 on Build GRCm38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erc2 | NM_177814.4 | NP_808482.2 | ||
| AK051416.1 | BAC34632.2 | |||
| AK122265.1 | BAC65547.1 | |||
| AK147213.1 | BAE27770.1 | |||
| BC056760.1 | AAH56760.1 |
More Information
Set Membership: |
|

Genomic Map