Assay Details
Target Gene Details
Entrez Gene ID: | 239037 |
Gene Name: | leucine-rich repeat, immunoglobulin-like and transmembrane domains 1 |
Gene Aliases: |
BC032270, Lrrc21 |
Location: |
Chr.14:37054830-37064938 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrit1 | NM_146245.2 | 3 | 789 | NP_666357.2 |
| AK044631.1 | 3 | 790 | BAC32010.1 | |
| BC032270.1 | 3 | 795 |
More Information
Set Membership: |
|

Genomic Map