Assay Details
Target Gene Details
Entrez Gene ID: | 75409 |
Gene Name: | SLIT and NTRK-like family, member 5 |
Gene Aliases: |
2610019D03Rik |
Location: |
Chr.14:111675115-111683144 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slitrk5 | NM_198865.1 | 2 | 3401 | NP_942565.1 |
| AK011457.2 | 1 | 300 | ||
| AK173062.1 | 1 | 2432 | BAD32340.1 | |
| BC065107.1 | 3 | 2736 | AAH65107.1 |

Genomic Map