Assay Details
Target Gene Details
Entrez Gene ID: | 239318 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 3 |
Gene Aliases: |
B130016O10Rik |
Location: |
Chr.15:4375491-4575585 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcxd3 | NM_177355.3 | 1 | 107 | NP_796329.2 |
| AK038575.1 | 1 | 107 | BAC30052.1 |

Genomic Map