Assay Details
Target Gene Details
Entrez Gene ID: | 68841 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma, opposite strand |
Gene Aliases: |
1110054M08Rik, AI481106 |
Location: |
Chr.16:24392556-24393655 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lppos | NR_037954.1 | 2 | 622 | |
AI481106.1 | 1 | 264 | ||
AK004252.1 | 2 | 410 |
