Assay Details
Target Gene Details
Entrez Gene ID: | 12300 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 2 |
Gene Aliases: |
AW060990, B230105C07Rik, B930041E13Rik, stargazer, stargazin, stg, wag, waggler |
Location: |
Chr.15:77993623-78119280 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng2 | NM_007583.2 | 4 | 2204 | NP_031609.1 |
| AK020958.1 | 1 | 553 | ||
| AK047154.1 | 1 | 350 |
More Information
Set Membership: |
|

Genomic Map