Assay Details
Target Gene Details
Entrez Gene ID: | 17444 |
Gene Name: | GRB2-related adaptor protein 2 |
Gene Aliases: |
GRAP-2, GRB2L, GRBLG, GRID, Gads, GrbX, Grf40, GrpL, Mona, P38 |
Location: |
Chr.15:80572422-80653047 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grap2 | NM_001289442.1 | 8 | 2202 | NP_001276371.1 |
| NM_010815.3 | 7 | 2297 | NP_034945.1 | |
| AF055465.1 | 7 | 2297 | AAD08803.1 | |
| AK030809.1 | 1 | 778 | ||
| AK156939.1 | 7 | 2276 | BAE33906.1 |

Genomic Map