Assay Details
Target Gene Details
Entrez Gene ID: | 17189 |
Gene Name: | myoglobin |
Gene Aliases: |
AI325109 |
Location: |
Chr.15:77015487-77050668 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mb | NM_001164047.1 | 3 | 915 | NP_001157519.1 |
NM_001164048.1 | 4 | 775 | NP_001157520.1 | |
NM_013593.3 | 4 | 863 | NP_038621.2 | |
AK137456.1 | 3 | 770 | BAE23359.1 | |
BC025172.1 | 4 | 875 | AAH25172.1 | |
BX527493.1 | 1 | 178 |
