Assay Details
Target Gene Details
Entrez Gene ID: | 239318 |
Gene Name: | phosphatidylinositol-specific phospholipase C, X domain containing 3 |
Gene Aliases: |
B130016O10Rik |
Location: |
Chr.15:4375491-4575585 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plcxd3 | NM_177355.3 | 2 | 438 | NP_796329.2 |
AK038575.1 | 3 | 529 | BAC30052.1 | |
AK044972.1 | 2 | 285 | BAC32164.1 | |
BC137637.1 | 2 | 293 | ||
BC137639.1 | 2 | 293 |
