Assay Details
Target Gene Details
Entrez Gene ID: | 12524 |
Gene Name: | CD86 antigen |
Gene Aliases: |
B7, B7-2, B7.2, B70, CLS1, Cd28l2, ETC-1, Ly-58, Ly58, MB7, MB7-2, TS/A-2 |
Location: |
Chr.16:36568956-36666077 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd86 | NM_019388.3 | 8 | 1316 | NP_062261.3 |
| AK079513.1 | 8 | 1317 | BAC37666.1 | |
| AK170054.1 | 8 | 1317 | BAE41535.1 | |
| BC013807.1 | 8 | 1290 | AAH13807.1 |
More Information
Set Membership: |
|

Genomic Map