Assay Details
Target Gene Details
Entrez Gene ID: | 14167 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
AV114868, B230343J05Rik, FGF-12, FHF-1, Fhf1 |
Location: |
Chr.16:28157783-28753243 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgf12 | AK034335.1 | 5 | 1871 | BAC28679.1 |
| BC030485.1 | 6 | 1835 | AAH30485.1 | |
| EU234007.1 | 3 | 1485 |

Genomic Map