Assay Details
Target Gene Details
Entrez Gene ID: | 66497 |
Gene Name: | cms small ribosomal subunit 1 |
Gene Aliases: |
1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629 |
Location: |
Chr.16:57302000-57606867 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmss1 | NM_025599.3 | NP_079875.2 | ||
| AK012168.1 | BAB28074.1 | |||
| AK019808.1 | BAB31859.2 | |||
| AK019979.1 | BAB31948.1 | |||
| BC038891.1 | AAH38891.1 |
Target Gene Details
Entrez Gene ID: | 78749 |
Gene Name: | filamin A interacting protein 1-like |
Gene Aliases: |
4631422O05Rik, Doc1 |
Location: |
Chr.16:57353277-57572804 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Filip1l | NM_001040397.4 | 2 | 264 | NP_001035487.2 |
| AK146705.1 | 2 | 265 | BAE27371.1 | |
| BY739488.1 | 2 | 265 | ||
| EU704258.1 | 2 | 111 |
More Information
Set Membership: |
|

Genomic Map