Assay Details
Target Gene Details
Entrez Gene ID: | 69572 |
Gene Name: | major facilitator superfamily domain containing 3 |
Gene Aliases: |
2310010G13Rik |
Location: |
Chr.15:76701542-76704239 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd3 | NM_027122.3 | 1 | 138 | NP_081398.2 |
| AK009269.1 | 1 | 217 | BAB26184.1 | |
| AK089539.1 | 1 | 139 | BAC40919.1 | |
| BC019171.1 | 1 | 165 | AAH19171.1 | |
| BC085305.1 | 1 | 202 | AAH85305.1 |
More Information
Set Membership: |
|

Genomic Map