Assay Details
Target Gene Details
Entrez Gene ID: | 71726 |
Gene Name: | single-strand selective monofunctional uracil DNA glycosylase |
Gene Aliases: |
1200013B09Rik, A930006H09Rik, C85220 |
Location: |
Chr.15:103153290-103168895 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smug1 | NM_027885.3 | 5 | 1668 | NP_082161.2 |
| AK004735.1 | 4 | 1404 | BAB23517.1 | |
| AK020817.1 | ||||
| AK045411.1 | 2 | 2743 | ||
| AK172510.1 | 4 | 1401 | BAE43042.1 | |
| BC050253.1 | 8 | 1944 | AAH50253.1 | |
| BC062960.1 | 5 | 1668 | AAH62960.1 |
More Information
Set Membership: |
|

Genomic Map