Assay Details
Target Gene Details
Entrez Gene ID: | 20665 |
Gene Name: | SRY (sex determining region Y)-box 10 |
Gene Aliases: |
Dom, Sox21 |
Location: |
Chr.15:79154909-79165235 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sox10 | NM_011437.1 | 2 | 292 | NP_035567.1 |
AF017182.1 | 3 | 354 | AAB99738.1 | |
AF047043.1 | AAC24564.1 | |||
AF047389.1 | AAC24565.1 | |||
AK042542.1 | 2 | 293 | ||
AK043220.1 | 2 | 289 | BAC31498.1 | |
AK185165.1 | ||||
AK185297.1 | ||||
AK193670.1 | ||||
BC018551.1 | 1 | 90 | AAH18551.1 | |
BC023356.1 | 2 | 126 | AAH23356.1 | |
BC025171.1 | 2 | 255 | AAH25171.1 | |
BC051058.1 | 2 | 126 | AAH51058.1 | |
U66141.1 | 2 | 290 | AAB49282.1 |
