Assay Details
Target Gene Details
Entrez Gene ID: | 12609 |
Gene Name: | CCAAT/enhancer binding protein (C/EBP), delta |
Gene Aliases: |
- |
Location: |
Chr.16:15887286-15889545 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cebpd | NM_007679.4 | 1 | 1872 | NP_031705.3 |
| AK053190.1 | 1 | 1872 | BAC35305.1 |
More Information
Set Membership: |
|

Genomic Map