Assay Details
Target Gene Details
Entrez Gene ID: | 14432 |
Gene Name: | growth associated protein 43 |
Gene Aliases: |
B-50, Basp2, GAP-43 |
Location: |
Chr.16:42248552-42340651 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gap43 | NM_008083.2 | 2 | 282 | NP_032109.1 |
| AK031908.1 | 2 | 185 | ||
| BC028288.1 | 2 | 282 | AAH28288.1 | |
| BC080758.1 | 2 | 197 | AAH80758.1 | |
| J02809.1 | 2 | 276 | AAA37377.1 |

Genomic Map