Assay Details
Target Gene Details
| Entrez Gene ID: | 380993 | 
| Gene Name: | zinc finger and AT hook domain containing | 
| Gene Aliases: | Gm922, Zfat1, Zfp406 | 
| Location: | Chr.15:68083714-68259265 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Zfat | NM_001145888.1 | 6 | 1437 | NP_001139360.1 | 
| NM_198644.2 | 6 | 1416 | NP_941046.1 | |
| BC053930.1 | 6 | 1407 | AAH53930.1 | |
| EU221277.1 | 6 | 1267 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top