Assay Details
Target Gene Details
Entrez Gene ID: | 70713 |
Gene Name: | G protein-coupled receptor 137C |
Gene Aliases: |
6330416L11Rik, Gm908, TM7SF1L2 |
Location: |
Chr.14:45219686-45282735 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpr137c | NM_027518.2 | 7 | 1720 | NP_081794.2 |
