Assay Details
Target Gene Details
Entrez Gene ID: | 18155 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, N23K, Npnc1, OFQ/N |
Location: |
Chr.14:65400673-65425581 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnoc | NM_001205075.1 | NP_001192004.1 | ||
| NM_010932.2 | 3 | 886 | NP_035062.1 | |
| AK045893.1 | 3 | 623 | BAC32523.1 | |
| BC119372.1 | 3 | 574 | ||
| BC144865.1 | ||||
| D50055.1 | BAA08773.1 | |||
| D50056.1 | 3 | 583 | BAA08774.1 | |
| D82866.1 | 3 | 886 | BAA11614.1 | |
| KF006993.1 | ||||
| U44027.1 | 1 | 294 | AAC52581.1 | |
| X97373.1 | 2 | 551 | CAA66042.1 |
More Information
Set Membership: |
|

Genomic Map