Assay Details
Target Gene Details
Entrez Gene ID: | 105675 |
Gene Name: | peptidylprolyl isomerase F (cyclophilin F) |
Gene Aliases: |
AW457192, CyP-D, CyP-F, CypD, PPIase |
Location: |
Chr.14:25693574-25700468 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppif | NM_134084.1 | 6 | 1217 | NP_598845.1 |
| AK028349.1 | 4 | 3310 | ||
| AK186554.1 | 1 | 295 | ||
| AK188218.1 | 1 | 322 | ||
| BC004041.1 | 6 | 1217 | AAH04041.1 |
Target Gene Details
Entrez Gene ID: | 68173 |
Gene Name: | peptidylprolyl isomerase F, opposite strand |
Gene Aliases: |
4930520F12Rik, Ppifs |
Location: |
Chr.14:25696356-25701282 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppifos | NR_028021.1 | |||
| AK015792.1 | ||||
| AK019686.1 | BAB31835.1 |
More Information
Set Membership: |
|

Genomic Map