Assay Details
Target Gene Details
Entrez Gene ID: | 66231 |
Gene Name: | THO complex 7 homolog (Drosophila) |
Gene Aliases: |
Nif3l1bp1 |
Location: |
Chr.14:13949012-13962042 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thoc7 | NM_001285780.1 | NP_001272709.1 | ||
| NM_025435.3 | NP_079711.1 | |||
| AK005176.1 | BAB23863.1 | |||
| AK007440.1 | BAB25040.1 | |||
| AK010958.1 | BAB27291.1 | |||
| AK013319.1 | 5 | 505 | ||
| AK143106.1 | 2 | 3542 | ||
| AK147003.1 | BAE27600.1 | |||
| BC042209.1 | AAH42209.1 | |||
| BC054419.1 | AAH54419.1 | |||
| BC116919.1 | ||||
| BC116945.1 | ||||
| CK329337.1 |
More Information
Set Membership: |
|

Genomic Map