Assay Details
Target Gene Details
Entrez Gene ID: | 66231 |
Gene Name: | THO complex 7 homolog (Drosophila) |
Gene Aliases: |
Nif3l1bp1 |
Location: |
Chr.14:13949012-13962042 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Thoc7 | NM_001285780.1 | 9 | 792 | NP_001272709.1 |
| NM_025435.3 | 8 | 714 | NP_079711.1 | |
| AK005176.1 | 8 | 641 | BAB23863.1 | |
| AK007440.1 | 8 | 620 | BAB25040.1 | |
| AK010958.1 | 8 | 655 | BAB27291.1 | |
| AK147003.1 | 9 | 695 | BAE27600.1 | |
| AK214063.1 | 2 | 104 | ||
| BC042209.1 | 8 | 641 | AAH42209.1 | |
| BC054419.1 | 8 | 674 | AAH54419.1 | |
| BC116919.1 | ||||
| BC116945.1 | ||||
| CF949442.1 | 3 | 186 |

Genomic Map