Assay Details
Target Gene Details
Entrez Gene ID: | 320865 |
Gene Name: | cadherin 18 |
Gene Aliases: |
B230220E17Rik, Cdh14l |
Location: |
Chr.15:22548899-23474432 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh18 | NM_001081299.1 | 2 | 354 | NP_001074768.1 |
| AK045672.1 | 2 | 506 | BAC32451.1 | |
| AK082311.1 | 2 | 363 | ||
| AK137430.1 | 2 | 354 |
More Information
Set Membership: |
|

Genomic Map