Assay Details
Target Gene Details
Entrez Gene ID: | 12563 |
Gene Name: | cadherin 6 |
Gene Aliases: |
K-cadherin, cad6, cadherin-6 |
Location: |
Chr.15:13028699-13173689 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh6 | NM_007666.3 | 2 | 191 | NP_031692.2 |
| AK134611.1 | 2 | 228 | BAE22207.1 | |
| CN704843.1 | 2 | 191 |
More Information
Set Membership: |
|

Genomic Map