Assay Details
Target Gene Details
Entrez Gene ID: | 56274 |
Gene Name: | serine/threonine kinase 3 |
Gene Aliases: |
0610042I06Rik, MST, Mst2, Mst3, Ste20, mess1 |
Location: |
Chr.15:34875496-35155806 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stk3 | NM_019635.2 | 11 | 2252 | NP_062609.2 |
| AK002914.1 | 6 | 1470 | BAB22453.1 | |
| AK004141.1 | 2 | 828 | ||
| AY058922.1 | 11 | 2138 | AAL29682.1 | |
| BC037440.1 | 9 | 2008 | AAH37440.1 | |
| BC049123.1 | 11 | 2126 | AAH49123.2 |
More Information
Set Membership: |
|

Genomic Map