Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gmpr2 |
Target Gene Details
Entrez Gene ID: | 28113 |
Gene Name: | Terf1 (TRF1)-interacting nuclear factor 2 |
Gene Aliases: |
AW552114, D14Wsu146e, Tin2 |
Location: |
Chr.14:55676208-55682624 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tinf2 | NM_145705.3 | 9 | 1466 | NP_663751.2 |
AF214013.1 | 9 | 1454 | AAF23504.1 | |
AF518764.1 | 9 | 1431 | AAN32986.1 | |
AI844697.1 | 1 | 274 | ||
AK011154.1 | 9 | 1434 | ||
AK016456.1 | 9 | 1400 | ||
AK028952.1 | 8 | 1591 | ||
AK090324.1 | 9 | 1433 | ||
AK144592.1 | BAE25954.1 | |||
AK169480.1 | 3 | 1583 | ||
AK205383.1 | 2 | 130 | ||
AK205420.1 | 2 | 127 | ||
AK214714.1 | 3 | 173 | ||
BC017142.1 | 4 | 1882 | ||
BC030347.1 | 9 | 1392 | AAH30347.1 |
