Assay Details
Target Gene Details
Entrez Gene ID: | 14114 |
Gene Name: | fibulin 1 |
Gene Aliases: |
- |
Location: |
Chr.15:85205944-85286294 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln1 | NM_010180.2 | 17 | 2724 | NP_034310.2 |
AK085989.1 | 17 | 2667 | ||
AK137511.1 | 17 | 2671 | ||
AK159646.1 | BAE35258.1 | |||
AK159816.1 | BAE35396.1 | |||
X70854.1 | CAA50207.1 |
