Assay Details
Target Gene Details
Entrez Gene ID: | 67154 |
Gene Name: | metadherin |
Gene Aliases: |
2610103J23Rik, 3D3, 3D3/Lyric, AEG-1, AV353288, D8Bwg1112e, Lyric |
Location: |
Chr.15:34082719-34143537 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtdh | NM_026002.4 | 1 | 229 | NP_080278.3 |
AF501309.1 | 1 | 167 | AAP30790.1 | |
AK009253.1 | 1 | 143 | ||
AK011808.1 | 1 | 224 | BAB27854.1 | |
AK029915.1 | 1 | 230 | BAC26673.1 | |
AK087486.1 | 1 | 255 | ||
AK151269.1 | 1 | 213 | BAE30256.1 | |
AK151810.1 | 1 | 224 | BAE30709.1 | |
AK156119.1 | 1 | 233 | ||
AK171976.1 | 1 | 221 | BAE42756.1 | |
BC023115.1 | 1 | 128 |
