Assay Details
Target Gene Details
Entrez Gene ID: | 67154 |
Gene Name: | metadherin |
Gene Aliases: |
2610103J23Rik, 3D3, 3D3/Lyric, AEG-1, AV353288, D8Bwg1112e, Lyric |
Location: |
Chr.15:34082719-34143537 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtdh | NM_026002.4 | 12 | 2836 | NP_080278.3 |
| AK034836.1 | 1 | 188 | ||
| AK035302.1 | 4 | 1230 | BAC29022.1 | |
| AK151269.1 | 12 | 2820 | BAE30256.1 | |
| AK151810.1 | 12 | 2831 | BAE30709.1 | |
| BC138858.1 | 12 | 2550 | ||
| BC138859.1 | 12 | 2550 |

Genomic Map