Assay Details
Target Gene Details
Entrez Gene ID: | 19354 |
Gene Name: | RAS-related C3 botulinum substrate 2 |
Gene Aliases: |
AI323801, AI452260 |
Location: |
Chr.15:78559169-78572783 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rac2 | NM_009008.3 | 7 | 1108 | NP_033034.1 |
| AK007561.1 | 7 | 1046 | BAB25109.1 | |
| AK011045.1 | 7 | 1090 | BAC25321.1 | |
| AK088415.1 | 7 | 1057 | ||
| AK088805.1 | 7 | 1045 | ||
| AK144136.1 | 7 | 1052 | BAE25721.1 | |
| AK171321.1 | 7 | 1109 | BAE42390.1 | |
| BC005455.1 | 7 | 1024 | AAH05455.1 | |
| X53247.1 | 7 | 1006 | CAA37337.1 |
More Information
Set Membership: |
|

Genomic Map