Assay Details
Target Gene Details
Entrez Gene ID: | 19214 |
Gene Name: | prostaglandin D receptor |
Gene Aliases: |
DP, PGD |
Location: |
Chr.14:44851235-44859444 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptgdr | NM_008962.4 | 1 | 168 | NP_032988.3 |
| AK032488.1 | 1 | 169 | BAC27895.1 | |
| BC132297.1 | 1 | 147 |
More Information
Set Membership: |
|

Genomic Map