Assay Details
Target Gene Details
Entrez Gene ID: | 57254 |
Gene Name: | taste receptor, type 2, member 119 |
Gene Aliases: |
T2r19, Tas2r19, mGR19, mt2r19 |
Location: |
Chr.15:32177289-32178294 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r119 | NM_020503.2 | 1 | 332 | NP_065249.2 |
| BC111811.1 | 1 | 333 | AAI11812.1 | |
| BC128078.1 | 1 | 329 | ||
| BC128079.1 | 1 | 333 | ||
| BC152359.1 | 1 | 333 |
More Information
Set Membership: |
|

Genomic Map