Assay Details
Target Gene Details
Entrez Gene ID: | 387609 |
Gene Name: | zinc fingers and homeoboxes 2 |
Gene Aliases: |
Afr-1, Afr1, Raf, mKIAA0854 |
Location: |
Chr.15:57694667-57839832 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zhx2 | NM_199449.2 | 4 | 3749 | NP_955520.1 |
| AK031782.1 | 4 | 3646 | BAC27546.1 | |
| AK157631.1 | 4 | 3736 | BAE34139.1 | |
| BC059178.1 | 4 | 3503 | AAH59178.1 | |
| BQ174722.1 |
More Information
Set Membership: |
|

Genomic Map