Assay Details
Target Gene Details
| Entrez Gene ID: | 13052 | 
| Gene Name: | coxsackie virus and adenovirus receptor | 
| Gene Aliases: | 2610206D03Rik, AU016810, AW553441, CAR, MCAR, MCVADR | 
| Location: | Chr.16:78301460-78359785 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 7 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Cxadr | NM_001025192.3 | 7 | 4794 | NP_001020363.1 | 
| NM_009988.4 | NP_034118.1 | |||
| AK011900.1 | 1 | 657 | ||
| AK141274.1 | 1 | 1684 | ||
| AK160457.1 | BAE35798.1 | |||
| BC016457.1 | AAH16457.1 | |||
| U90715.1 | AAC53148.1 | |||
| Y11929.1 | CAA72679.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic