Assay Details
Target Gene Details
Entrez Gene ID: | 13052 |
Gene Name: | coxsackie virus and adenovirus receptor |
Gene Aliases: |
2610206D03Rik, AU016810, AW553441, CAR, MCAR, MCVADR |
Location: |
Chr.16:78301460-78359785 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxadr | NM_009988.4 | 8 | 1646 | NP_034118.1 |
| BC016457.1 | 8 | 1561 | AAH16457.1 | |
| Y11929.1 | 8 | 1600 | CAA72679.1 |

Genomic Map