Assay Details
Target Gene Details
Entrez Gene ID: | 11732 |
Gene Name: | progressive ankylosis |
Gene Aliases: |
Ankh, D15Ertd221e, ank, mKIAA1581 |
Location: |
Chr.15:27466677-27594909 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ank | NM_020332.4 | 12 | 1993 | NP_065065.3 |
AF001533.2 | 10 | 1339 | AAB65654.2 | |
AF274752.1 | 12 | 1696 | AAF88038.1 | |
AK083135.1 | 12 | 1994 | BAC38778.1 | |
AK086014.1 | 5 | 864 | ||
AK148070.1 | 12 | 1766 | BAE28324.1 | |
BC054379.1 | 12 | 1900 | AAH54379.1 | |
DQ832285.1 | 12 | 1993 |
