Assay Details
Target Gene Details
Entrez Gene ID: | 93686 |
Gene Name: | RNA binding protein, fox-1 homolog (C. elegans) 2 |
Gene Aliases: |
Fbm2, Fxh, Hrnbp2, Rbm9 |
Location: |
Chr.15:77078990-77307053 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbfox2 | NM_001110827.2 | 14 | 5281 | NP_001104297.1 |
| NM_001110828.2 | 13 | 4835 | NP_001104298.1 | |
| NM_001110829.2 | 13 | 4826 | NP_001104299.1 | |
| NM_001110830.2 | 13 | 4823 | NP_001104300.1 | |
| NM_001286417.1 | 12 | 2848 | NP_001273346.1 | |
| NM_001286418.1 | 10 | 4541 | NP_001273347.1 | |
| NM_001286419.1 | 12 | 4730 | NP_001273348.1 | |
| NM_053104.6 | 14 | 5293 | NP_444334.3 | |
| NM_175387.3 | 13 | 5241 | NP_780596.1 | |
| AK044929.1 | 12 | 2823 | BAC32147.1 |
More Information
Set Membership: |
|

Genomic Map