Assay Details
Target Gene Details
Entrez Gene ID: | 75766 |
Gene Name: | dentrocyte expressed seven transmembrane protein |
Gene Aliases: |
4833414I07Rik, DC-STAMP, FIND, Tm7sf4, mDC-STAMP |
Location: |
Chr.15:39745930-39760938 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dcstamp | NM_001289506.1 | 2 | 735 | NP_001276435.1 |
NM_001289508.1 | 2 | 735 | NP_001276437.1 | |
NM_001289512.1 | 2 | 735 | NP_001276441.1 | |
NM_001289513.1 | 2 | 735 | NP_001276442.1 | |
NM_029422.4 | 2 | 735 | NP_083698.1 | |
AB109560.1 | 2 | 733 | BAC81438.1 | |
AB109561.1 | 2 | 733 | BAC81439.1 | |
AK014697.1 | 2 | 734 | BAB29508.1 | |
AK154384.1 | 2 | 734 | BAE32551.1 | |
AY517483.1 | 2 | 734 | AAR99406.1 | |
AY517484.1 | 3 | 485 | AAR99407.1 | |
AY517485.1 | 2 | 734 | AAR99408.1 | |
BC139146.1 | 2 | 721 | ||
BC145558.1 | 2 | 721 |
