Assay Details
Target Gene Details
Entrez Gene ID: | 71000 |
Gene Name: | RIKEN cDNA 4931440J10 gene |
Gene Aliases: |
- |
Location: |
Chr.14:62760501-62830126 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931440J10Rik | NR_045503.1 | |||
| AK016511.1 |
Target Gene Details
Entrez Gene ID: | 18130 |
Gene Name: | integrator complex subunit 6 |
Gene Aliases: |
2900075H24Rik, AI480962, DICE1, Ddx26, HDB, Notch2l |
Location: |
Chr.14:62676325-62761370 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ints6 | NM_008715.2 | 1 | 480 | NP_032741.2 |
| AK053385.1 | 1 | 502 | ||
| AK149711.1 | 1 | 507 | ||
| AK172178.1 | 1 | 508 | BAE42868.1 | |
| BC059263.1 | 1 | 480 | AAH59263.1 |
More Information
Set Membership: |
|

Genomic Map